missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAA1513422
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1513422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1513422 Supplier Thermo Scientific Chemicals Supplier No. A1513422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

2-Chloro-N,N-dimethylethylamine is a widely used as a intermediate and starting reagent for organic synthesis. Also acts as Intermediate for the syntheses of bephenium hydroxynaphthoate, diltiazem, mepyramine and phenyltoloxamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N,N-dimethylethylamine is a widely used as a intermediate and starting reagent for organic synthesis. Also acts as Intermediate for the syntheses of bephenium hydroxynaphthoate, diltiazem, mepyramine and phenyltoloxamine.

Chemical Identifiers

CAS 4584-46-7
Molecular Formula C4H11Cl2N
Molecular Weight (g/mol) 144.039
MDL Number MFCD00012516
InChI Key LQLJZSJKRYTKTP-UHFFFAOYSA-N
Synonym 2-dimethylaminoethyl chloride hydrochloride, 2-chloro-n,n-dimethylethanamine hydrochloride, 2-dimethylamino ethyl chloride hydrochloride, 2-chloroethyl dimethylamine hydrochloride, 2-chloro-n,n-dimethylethylamine hydrochloride, dimethylaminoethyl chloride hydrochloride, 2-chloroethyldimethylamine hydrochloride, 1-chloro-2-dimethylaminoethane hydrochloride, 2-dimethylamino ethylchloride hydrochloride, n-2-chloroethyl dimethylamine hydrochloride
PubChem CID 11636816
ChEBI CHEBI:78153
IUPAC Name 2-chloro-N,N-dimethylethanamine;hydrochloride
SMILES CN(C)CCCl.Cl

Specifications

Melting Point 205°C to 208°C
Quantity 100 g
UN Number UN2811
Beilstein 3591234
Sensitivity Hygroscopic
Solubility Information Soluble in water (2000mg/ml at 20°C),and alcohol.
Formula Weight 144.05
Percent Purity ≥98%
Chemical Name or Material 2-Dimethylaminoethyl chloride hydrochloride
Hazard Category H301-H311-H315-H320-H335-H341-H372
Hazard Statement GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 224-970-1
RTECSNumber KQ9020000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only