missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoethyl Benzoate 96.0+%, TCI America™

Catalog No. B126625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B126625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B126625G Supplier TCI America Supplier No. B126625G
Only null left

Chemical Identifiers

CAS 2208-05-1
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD00051067
InChI Key KJSGODDTWRXQRH-UHFFFAOYSA-N
Synonym Benzoic Acid 2-Dimethylaminoethyl Ester
PubChem CID 75158
IUPAC Name 2-(dimethylamino)ethyl benzoate
SMILES CN(C)CCOC(=O)C1=CC=CC=C1

Specifications

Boiling Point 159°C
Quantity 25 g
Formula Weight 193.25
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Dimethylaminoethyl Benzoate