missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)pyridine, 97%

Catalog No. aal0251314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0251314 25 g
AAL0251306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0251314 Supplier Thermo Scientific Chemicals Supplier No. L0251314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5683-33-0 | C7H10N2 | 122.17 g/mol

2-(Dimethylamino)pyridine in toluene at room temperature in the presence of 1.1 equivalent of methyl trifluoromethanesulfonate forms (2-pyridyl)-trimethylammonium trifluoromethanesulfonate salt. It is used as a model substrate in the preparation of chelate carbene via cyclometalation, H2 loss and reversible α-elimination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)pyridine in toluene at room temperature in the presence of 1.1 equivalent of methyl trifluoromethanesulfonate forms (2-pyridyl)-trimethylammonium trifluoromethanesulfonate salt. It is used as a model substrate in the preparation of chelate carbene via cyclometalation, H2 loss and reversible α-elimination.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5683-33-0
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00006261
InChI Key PSHKMPUSSFXUIA-UHFFFAOYSA-N
Synonym 2-dimethylaminopyridine, 2-dimethylaminopyridin, dimethylaminopyridine, 2-dimethylamino pyridine, 2-dimethylaminopyridine cobalt complex, 2-pyridinamine, n,n-dimethyl, dimethylamino-2 pyridine, dimethylaminopyridin, unii-18q95u3z8h, pyridine, 2-dimethylamino
PubChem CID 21885
IUPAC Name N,N-dimethylpyridin-2-amine
SMILES CN(C)C1=CC=CC=N1

Specifications

Density 1.008
Boiling Point 190°C to 192°C
Flash Point 80°C (176°F)
Refractive Index 1.559
Quantity 25 g
Beilstein 110353
Solubility Information Soluble in water.
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 2-(Dimethylamino)pyridine
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 227-147-5
RTECSNumber US9200000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only