missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. p-7117323
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A123525G 25 g
A1235100G 100 g
A1235500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A123525G Supplier TCI America Supplier No. A123525G
Only null left

Chemical Identifiers

CAS 2439-35-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00038233
InChI Key DPBJAVGHACCNRL-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl acrylate, dimethylaminoethyl acrylate, n,n-dimethylaminoethyl acrylate, adame, 2-propenoic acid, 2-dimethylamino ethyl ester, acrylic acid, 2-dimethylamino ethyl ester, 2-dimethylamino, unii-5yes121ftl, 2-n,n-dimethylamino ethyl acrylate, ageflex fa-1
PubChem CID 17111
IUPAC Name 2-(dimethylamino)ethyl prop-2-enoate
SMILES CN(C)CCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 161°C
Quantity 25 g
UN Number 3302
Formula Weight 143.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Dimethylamino)ethyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-2583
RTECSNumber AS8578000
TSCA Yes
Recommended Storage Refrigerator