missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)-2-methyl-1-propanol 97.0+%, TCI America™

Catalog No. D109825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D109825ML 25 mL
D1098500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D109825ML Supplier TCI America Supplier No. D109825ML

Chemical Identifiers

CAS 7005-47-2
Molecular Formula C6H16ClNO
Molecular Weight (g/mol) 153.65
MDL Number MFCD00021978
InChI Key QVNXEWQCNPSJLG-UHFFFAOYSA-N
Synonym dmamp, 2-dimethylamino-2-methylpropan-1-ol, 1-propanol, 2-dimethylamino-2-methyl, usaf cs-1, 2-dimethylamino-2-methyl-1-propanol, 2-dimethylamino-2-methylpropanol, unii-6zwg40uq9e, 2-n,n-dimethylamino isobutanol, 6zwg40uq9e
PubChem CID 23435
IUPAC Name hydrogen 2-(dimethylamino)-2-methylpropan-1-ol chloride
SMILES [H+].[Cl-].CN(C)C(C)(C)CO

Specifications

Color Yellow
Boiling Point 163°C
Quantity 25 mL
UN Number 2734
Formula Weight 117.19
Percent Purity ≥97.0% (GC,T)
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-(Dimethylamino)-2-methyl-1-propanol
RTECSNumber UB3325000
TSCA Yes