missing translation for 'onlineSavingsMsg'
Learn More

2-(Diisopropylsilyl)pyridine 95.0+%, TCI America™

Catalog No. p-7128185
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42641G 1 g
D42645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42641G Supplier TCI America Supplier No. D42641G

Chemical Identifiers

CAS 1232692-92-0
Molecular Formula C11H18NSi
Molecular Weight (g/mol) 192.36
MDL Number MFCD22200515
InChI Key MARDZIUPIJQZBI-UHFFFAOYSA-N
Synonym Diisopropyl(2-pyridyl)silane, PyDipSiH
PubChem CID 91972087
IUPAC Name bis(propan-2-yl)(pyridin-2-yl)silyl
SMILES CC(C)[Si](C(C)C)C1=CC=CC=N1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 193.37
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Diisopropylsilyl)pyridine