missing translation for 'onlineSavingsMsg'
Learn More

2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%

Catalog No. AAA1748518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1748518 50 g
AAA1748530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1748518 Supplier Thermo Scientific Chemicals Supplier No. A1748518
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 51-05-8 | C13H21ClN2O2 | 272.77 g/mol

A sodium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51-05-8
Molecular Formula C13H21ClN2O2
Molecular Weight (g/mol) 272.77
MDL Number MFCD00013000
InChI Key HCBIBCJNVBAKAB-UHFFFAOYSA-N
Synonym procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
PubChem CID 5795
ChEBI CHEBI:8431
SMILES [H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1

Specifications

Melting Point 155°C to 157°C
Boiling Point 195°C to 196°C (17 mmHg)
Odor Odorless
Quantity 50 g
UN Number UN2811
Sensitivity Light sensitive
Merck Index 14,7757
IUPAC Name 2-(diethylamino)ethyl 4-aminobenzoate;hydrochloride
Formula Weight 272.78
Percent Purity 99%
Chemical Name or Material 2-Diethylaminoethyl 4-aminobenzoate hydrochloride
Show More Show Less
Hazard Category H301-H315-H317-H318-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-077-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only