missing translation for 'onlineSavingsMsg'
Learn More

2-Diethylaminoethyl-3-(1-naphthyl)-2-(1-naphthylmethyl)propionate oxalate, MilliporeSigma™ Supelco™

Catalog No. 111022892
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022892 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022892 Supplier Merck Emd Millipore Supplier No. BP361

British Pharmacopoeia (BP) Reference Standard

  • This product is provided as delivered and specified by the issuing Pharmacopoeia
  • All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia

Specifications

CAS 115025-99-5
Molecular Formula C30H33NO2 · C2H2O4
Linear Formula C30H33NO2 · C2H2O4
Quantity 25 mg
Synonym 2-Diethylaminoethyl 3-(1-naphthyl)-2-(1-naphthylmethyl)propionate oxalate salt
Formula Weight 529.62
Grade BP