missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Diethylamino)ethyl Bromide Hydrobromide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 208°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 261.00 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(Diethylamino)ethyl Bromide Hydrobromide |
Safety and Handling
| TSCA | No |