missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethyl Bromide Hydrobromide 98.0+%, TCI America™

Catalog No. D3232500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3232500G 500 g
D323225G 25 g
D3232100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D3232500G Supplier TCI America Supplier No. D3232500G

Chemical Identifiers

CAS 1069-72-3
Molecular Formula C6H15Br2N
Molecular Weight (g/mol) 261.00
MDL Number MFCD00040376
InChI Key HLMHCDKXKXBKQK-UHFFFAOYSA-N
Synonym 2-diethylamino ethyl bromide hydrobromide, 2-bromo-n,n-diethylethylamine hydrobromide, 2-bromoethyldiethylammonium bromide, 2-bromoethyl diethylamine hydrobromide, 2-bromo-n,n-diethylethanaminium bromide, 2-bromotriethylamine hydrobromide, 2-bromoethyldiethylamine hydrobromide, diethylaminoethyl bromide hydrobromide, 2-bromo-n,n-diethylethanamine hydrobromide, ethanamine, 2-bromo-n,n-diethyl-, hydrobromide
PubChem CID 70607
IUPAC Name hydrogen (2-bromoethyl)diethylamine bromide
SMILES [H+].[Br-].CCN(CC)CCBr

Specifications

Melting Point 208°C
Color White-Yellow
Quantity 500 g
Formula Weight 261.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Diethylamino)ethyl Bromide Hydrobromide