missing translation for 'onlineSavingsMsg'
Learn More

2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl 98.0+%, TCI America™

Catalog No. d33891g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33891G 1 g
D33895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33891G Supplier TCI America Supplier No. D33891G
Only null left

Chemical Identifiers

CAS 213697-53-1
Molecular Formula C26H36NP
Molecular Weight (g/mol) 393.555
InChI Key ZEMZPXWZVTUONV-UHFFFAOYSA-N
Synonym 2-dicyclohexylphosphino-2'-n,n-dimethylamino biphenyl, davephos, 2'-dicyclohexylphosphino-n,n-dimethyl-1,1'-biphenyl-2-amine, 2'-dicyclohexylphosphino-n,n-dimethylbiphenyl-2-amine, 2-dicyclohexylphosphino-2'-dimethylamino biphenyl, unii-9877onu78f, 2-2-dicyclohexylphosphanyl phenyl-n,n-dimethylaniline, 2'-dicyclohexylphosphanyl-n,n-dimethyl-1,1'-biphenyl-2-amine
PubChem CID 2734217
IUPAC Name 2-(2-dicyclohexylphosphanylphenyl)-N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 1 g
Formula Weight 393.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Dicyclohexylphosphino)-2′-(dimethylamino)biphenyl