missing translation for 'onlineSavingsMsg'
Learn More

2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl 98.0+%, TCI America™

Catalog No. D50365G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50365G 5 g
D50361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50365G Supplier TCI America Supplier No. D50365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 657408-07-6
Molecular Formula C26H35O2P
Molecular Weight (g/mol) 410.54
MDL Number MFCD05861611
InChI Key VNFWTIYUKDMAOP-UHFFFAOYSA-N
Synonym 2-dicyclohexylphosphino-2',6'-dimethoxybiphenyl, s-phos, sphos, dicyclohexyl 2',6'-dimethoxy-1,1'-biphenyl-2-yl phosphine, 2-dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl, unii-xi1mq32186, dicyclohexyl 2-2,6-dimethoxyphenyl phenyl phosphane, dicyclohexyl 2',6'-dimethoxybiphenyl-2-yl phosphine, 2-dicyclohexylphosphino-2,6-dimethoxy-1,1-biphenyl, 2-dicyclohexylphosphino-2',6-dimethoxybiphenyl
PubChem CID 11269872
IUPAC Name dicyclohexyl({2',6'-dimethoxy-[1,1'-biphenyl]-2-yl})phosphane
SMILES COC1=CC=CC(OC)=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 5 g
Formula Weight 410.54
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Dicyclohexylphosphino-2′,6′-dimethoxybiphenyl