missing translation for 'onlineSavingsMsg'
Learn More

2-Dicyclohexylphosphino-2',6'-diisopropoxybiphenyl 98.0+%, TCI America™

Catalog No. p-7128625
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50371G 1 g
D50375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50371G Supplier TCI America Supplier No. D50371G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 787618-22-8
Molecular Formula C30H43O2P
Molecular Weight (g/mol) 466.65
MDL Number MFCD06798294
InChI Key MXFYYFVVIIWKFE-UHFFFAOYSA-N
Synonym ruphos, dicyclohexyl 2',6'-diisopropoxy-1,1'-biphenyl-2-yl phosphine, 2-dicyclohexylphosphino-2',6'-diisopropoxy-1,1'-biphenyl, 2-dicyclohexylphosphino-2',6'-diisopropoxybiphenyl, 2-dicyclohexylphosphino-2',6'-di-i-propoxy-1,1'-biphenyl, unii-x3474ry19e, 2-dicyclohexylphosphino-2',6'-isopropoxybiphenyl, phosphine, 2',6'-bis 1-methylethoxy 1,1'-biphenyl-2-yl dicyclohexyl, 2-dicyclohexylphosphino-2,6-diisopropoxy-1,1-biphenyl, dicyclohexyl 2',6'-diisopropoxybiphenyl-2-yl phosphine
PubChem CID 16217985
IUPAC Name [2',6'-bis(propan-2-yloxy)-[1,1'-biphenyl]-2-yl]dicyclohexylphosphane
SMILES CC(C)OC1=CC=CC(OC(C)C)=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 125°C
Color White
Quantity 1 g
Formula Weight 466.65
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Dicyclohexylphosphino-2′,6′-diisopropoxybiphenyl