missing translation for 'onlineSavingsMsg'
Learn More

2-(Dibutylamino)ethanol 99.0+%, TCI America™

Catalog No. D0219500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0219500ML 500 mL
D021925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0219500ML Supplier TCI America Supplier No. D0219500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102-81-8
Molecular Formula C10H23NO
Molecular Weight (g/mol) 173.3
MDL Number MFCD00014033
InChI Key IWSZDQRGNFLMJS-UHFFFAOYSA-N
Synonym 2-dibutylamino ethanol, n,n-dibutylethanolamine, 2-n-dibutylaminoethanol, ethanol, 2-dibutylamino, dibutylethanolamine, di n-butyl ethanolamine, bu2ae, 2-di-n-butylaminoethanol, dbae, dibutylaminoethanol
PubChem CID 7621
IUPAC Name 2-(dibutylamino)ethanol
SMILES CCCCN(CCCC)CCO

Specifications

Melting Point -70°C
Color Yellow
Boiling Point 226°C
Quantity 500 mL
UN Number 2873
Formula Weight 173.30
Percent Purity ≥99.0% (GC)
Grade Not Available
Physical Form Liquid
Chemical Name or Material 2-(Dibutylamino)ethanol
EINECSNumber (2)-0353
RTECSNumber KK3850000
TSCA Yes