missing translation for 'onlineSavingsMsg'
Learn More

2-(Di-tert-butylphosphino)biphenyl 98.0+%, TCI America™

Catalog No. p-7127708
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33871G 1 g
D33875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33871G Supplier TCI America Supplier No. D33871G

Chemical Identifiers

CAS 224311-51-7
Molecular Formula C20H27P
Molecular Weight (g/mol) 298.41
MDL Number MFCD01862440
InChI Key CNXMDTWQWLGCPE-UHFFFAOYSA-N
Synonym 2-di-tert-butylphosphino biphenyl, johnphos, 2-di-t-butylphosphino biphenyl, biphenyl-2-yl-di-tert-butyl-phosphane, 2-biphenyl di-tert-butylphosphine, 2-biphenylyl di-tert-butylphosphine, 2-di-tert-butylphosphino-biphenyl, biphenyl-2-yldi-tert-butylphosphine, di-tert-butyl 2-phenylphenyl phosphane, 1,1'-biphenyl-2-yldi-tert-butylphosphine
PubChem CID 2734215
IUPAC Name ditert-butyl-(2-phenylphenyl)phosphane
SMILES CC(C)(C)P(C1=CC=CC=C1C2=CC=CC=C2)C(C)(C)C

Specifications

Melting Point 87°C
Color White
Quantity 1 g
Formula Weight 298.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Di-tert-butylphosphino)biphenyl