missing translation for 'onlineSavingsMsg'
Learn More

2-Di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl 98.0+%, TCI America™

$257.97 - $768.15

Chemical Identifiers

CAS 564483-19-8
Molecular Formula C29H45P
Molecular Weight (g/mol) 424.653
MDL Number MFCD06411306
InChI Key SACNIGZYDTUHKB-UHFFFAOYSA-N
Synonym 2-di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, di-tert-butyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, tbuxphos, tert-butyl xphos, t-butylxphos, 2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, t-bu xphos, di-t-bu-xphos, di-tert-butyl 2',4',6'-triisopropylbiphenyl-2-yl phosphine, 2-di-t-butylphosphino-2',4',6'-triisopropylbiphenyl
PubChem CID 11618717
IUPAC Name ditert-butyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D50391G
View Documents
TCI America
D50391G
1 g N/A
Only null left
 
D50395G
View Documents
TCI America
D50395G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

564483-19-8
424.653
SACNIGZYDTUHKB-UHFFFAOYSA-N
11618717
CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C)C(C)C
C29H45P
MFCD06411306
2-di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, di-tert-butyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, tbuxphos, tert-butyl xphos, t-butylxphos, 2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, t-bu xphos, di-t-bu-xphos, di-tert-butyl 2',4',6'-triisopropylbiphenyl-2-yl phosphine, 2-di-t-butylphosphino-2',4',6'-triisopropylbiphenyl
ditert-butyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane

Specifications

564483-19-8
White-Yellow
MFCD06411306
2-di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl, di-tert-butyl 2',4',6'-triisopropyl-1,1'-biphenyl-2-yl phosphine, tbuxphos, tert-butyl xphos, t-butylxphos, 2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl, t-bu xphos, di-t-bu-xphos, di-tert-butyl 2',4',6'-triisopropylbiphenyl-2-yl phosphine, 2-di-t-butylphosphino-2',4',6'-triisopropylbiphenyl
CC(C)C1=CC(=C(C(=C1)C(C)C)C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C)C(C)C
424.653
424.65
Crystalline Powder
148°C
C29H45P
1 g
SACNIGZYDTUHKB-UHFFFAOYSA-N
ditert-butyl-[2-[2,4,6-tri(propan-2-yl)phenyl]phenyl]phosphane
11618717
≥98.0% (HPLC,T)
2-Di-tert-butylphosphino-2′,4′,6′-triisopropylbiphenyl
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents