Learn More
2-[Di(methylthio)methylidene]malononitrile, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CD00793EA
Specifications
| 2-[Di(methylthio)methylidene]malononitrile | |
| 96.5 | |
| 97% | |
| C6H6N2S2 | |
| MFCD00052730 | |
| 2-bis methylthio methylene malononitrile, bis methylthio methylene malononitrile, 2-di methylthio methylidene malononitrile, propanedinitrile, bis methylthio methylene, 2-bis methylsulfanyl methylidene propanedinitrile, bis methylthio methylene propanedinitrile, 3,3-bis methylthio-2-cyanoacrylonitrile, bis methylthio methylidene propanedinitrile | |
| CSC(=C(C#N)C#N)SC | |
| 170.248 | |
| 170.26 |
| 5147-80-8 | |
| 100.0 | |
| Amber Glass Bottle | |
| (CN)2C=C(SCH3)2 | |
| 10 g | |
| FICQFRCPSFCFBY-UHFFFAOYSA-N | |
| 2-[bis(methylsulfanyl)methylidene]propanedinitrile | |
| 99229 | |
| 97% |
Chemical Identifiers
| 5147-80-8 | |
| 170.248 | |
| FICQFRCPSFCFBY-UHFFFAOYSA-N | |
| 99229 | |
| CSC(=C(C#N)C#N)SC |
| C6H6N2S2 | |
| MFCD00052730 | |
| 2-bis methylthio methylene malononitrile, bis methylthio methylene malononitrile, 2-di methylthio methylidene malononitrile, propanedinitrile, bis methylthio methylene, 2-bis methylsulfanyl methylidene propanedinitrile, bis methylthio methylene propanedinitrile, 3,3-bis methylthio-2-cyanoacrylonitrile, bis methylthio methylidene propanedinitrile | |
| 2-[bis(methylsulfanyl)methylidene]propanedinitrile |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger