missing translation for 'onlineSavingsMsg'
Learn More

2-[Di(methylthio)methylidene]malononitrile, 97%, Thermo Scientific™

Catalog No. CD00793DA
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CD00793DA 1 g Amber Glass Bottle
CD00793EA 10 g Amber Glass Bottle
CD00793EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CD00793DA Supplier Thermo Scientific Chemicals Supplier No. CD00793DA

Chemical Identifiers

CAS 5147-80-8
Molecular Formula C6H6N2S2
Molecular Weight (g/mol) 170.248
MDL Number MFCD00052730
InChI Key FICQFRCPSFCFBY-UHFFFAOYSA-N
Synonym 2-bis methylthio methylene malononitrile, bis methylthio methylene malononitrile, 2-di methylthio methylidene malononitrile, propanedinitrile, bis methylthio methylene, 2-bis methylsulfanyl methylidene propanedinitrile, bis methylthio methylene propanedinitrile, 3,3-bis methylthio-2-cyanoacrylonitrile, bis methylthio methylidene propanedinitrile
PubChem CID 99229
IUPAC Name 2-[bis(methylsulfanyl)methylidene]propanedinitrile
SMILES CSC(=C(C#N)C#N)SC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CN)2C=C(SCH3)2
Quantity 1 g
Formula Weight 170.26
Percent Purity 97%
Chemical Name or Material 2-[Di(methylthio)methylidene]malononitrile
Hazard Category Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.