missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Deoxyuridine 99.0+%, TCI America™
Supplier: TCI America D006025G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 951-78-0 | |
| 228.20 | |
| MXHRCPNRJAMMIM-SHYZEUOFSA-N | |
| 13712 | |
| 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| C9H12N2O5 | |
| MFCD00006527 | |
| 2'-deoxyuridine, deoxyuridine, uracil deoxyriboside, 2-deoxyuridine, 1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, deoxyribose uracil, uridine, 2'-deoxy, 2'-desoxyuridine, unii-w78i7ay22c, ccris 2832 | |
| CHEBI:16450 | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O |
Specifications
| 2′-Deoxyuridine | |
| 164°C | |
| C9H12N2O5 | |
| MFCD00006527 | |
| MXHRCPNRJAMMIM-SHYZEUOFSA-N | |
| 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 13712 | |
| 228.20 | |
| Crystalline Powder |
| 951-78-0 | |
| White | |
| 25 g | |
| 2'-deoxyuridine, deoxyuridine, uracil deoxyriboside, 2-deoxyuridine, 1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1h,3h-dione, deoxyribose uracil, uridine, 2'-deoxy, 2'-desoxyuridine, unii-w78i7ay22c, ccris 2832 | |
| OC[C@H]1O[C@H](C[C@@H]1O)N1C=CC(=O)NC1=O | |
| 228.20 | |
| CHEBI:16450 | |
| ≥99.0% (HPLC) |
Safety and Handling
EINECSNumber : (9)-2228
RTECSNumber : YU7490000
TSCA : Yes