missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyuridine-5'-triphosphate trisodium salt, 95%

Catalog No. aaj64716mb
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64716MB 25 mg
AAJ64716MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64716MB Supplier Thermo Scientific Chemicals Supplier No. J64716MB
Only null left

CAS: 102814-08-4 | C9H15N2Na3O14P3 | 537.11 g/mol

2'-Deoxyuridine-5'-triphosphate disodium salt is used in conjunction with uracil DNA glycosylase to prevent carryover contamination in PCR. The dUTP is substituted for dTTP in the PCR reaction, used to study the distribution, specificity and kinetics of deoxyuridine 5?-triphosphate nucleotidohydrolase(s) (dUTPase, EC 3.6.1.23). dUTPase are important enzymes that protect cells from the incorporation of dUTP into DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyuridine-5′-triphosphate disodium salt is used in conjunction with uracil DNA glycosylase to prevent carryover contamination in PCR. The dUTP is substituted for dTTP in the PCR reaction, used to study the distribution, specificity and kinetics of deoxyuridine 5′-triphosphate nucleotidohydrolase(s) (dUTPase, EC 3.6.1.23). dUTPase are important enzymes that protect cells from the incorporation of dUTP into DNA.

Solubility
Soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 102814-08-4
Molecular Formula C9H15N2Na3O14P3
Molecular Weight (g/mol) 537.11
MDL Number MFCD00084701
InChI Key AYFBRVJQNMGUSV-MILVPLDLSA-N
Synonym 2'-deoxyuridine-5'-triphosphate trisodium salt
PubChem CID 131843218
IUPAC Name [[(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium
SMILES C1C(C(OC1N2C=CC(=O)NC2=O)COP(=O)(O)OP(=O)(O)OP(=O)(O)O)O.[Na].[Na].[Na]

Specifications

Color White
Quantity 25 mg
Solubility Information Soluble in water.
Formula Weight 537.11
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2'-Deoxyuridine-5'-triphosphate disodium salt
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only