missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyuridine-5'-monophosphate disodium salt, 99%

CAS: 42155-08-8 | C9H13N2Na2O8P | 354.162 g/mol

Supplier:  Thermo Scientific Chemicals J64627MD

 View more versions of this product

Catalog No. AAJ64627MD


Only null left
Explore available promotions

Description

Description

2?-Deoxyuridine 5?-monophosphate (dUMP) is used by the enzyme thymidylate synthetase (TS) (EC 2.1.1.45) for the de novo production of dTMP. dUMP is used as the reference substrate in studies of dUMP analogues in potential chemotherapy applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyuridine 5′-monophosphate (dUMP) is used by the enzyme thymidylate synthetase (TS) (EC 2.1.1.45) for the de novo production of dTMP. dUMP is used as the reference substrate in studies of dUMP analogues in potential chemotherapy applications.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2'-Deoxyuridine-5'-monophosphate disodium salt
White
MFCD00065282
2'-deoxyuridine 5'-monophosphate disodium salt
CWTUMDATELIIAI-CDNBRZBRSA-N
[(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
129893468
99%
42155-08-8
C9H13N2Na2O8P
250 mg
Soluble in water.
C1C(C(OC1N2C=CC(=O)NC2=O)COP(=O)(O)O)O.[Na].[Na]
354.162
352.15
Powder

Chemical Identifiers

42155-08-8
354.162
CWTUMDATELIIAI-CDNBRZBRSA-N
129893468
C1C(C(OC1N2C=CC(=O)NC2=O)COP(=O)(O)O)O.[Na].[Na]
C9H13N2Na2O8P
MFCD00065282
2'-deoxyuridine 5'-monophosphate disodium salt
[(2R,3S,5R)-5-(2,4-dioxopyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 255-687-1

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only