Learn More
2'-Deoxyinosine-5'-monophosphate disodium salt, 99%, Thermo Scientific Chemicals
$175.48 - $175.48
Chemical Identifiers
| CAS | 14999-52-1 |
|---|---|
| Molecular Formula | C10H11N4Na2O7P |
| Molecular Weight (g/mol) | 376.17 |
| MDL Number | MFCD00134874 |
| InChI Key | VDPLPYMIJCLJEF-DZVPOJRENA-L |
| Synonym | sodium 2r,3s,5r-3-hydroxy-5-6-oxo-1h-purin-9 6h-yl tetrahydrofuran-2-yl methyl phosphate, 2'-deoxyinosine-5'-monophosphate, disodium salt, 2'-deoxyinosine 5'-monophosphate disodium salt, 2'-deoxyinosine-5'-monophosphate disodium salt, disodium dimp, 2'-deoxyinosine-5'-phosphate sodium, disodium 2'-deoxyinosine 5'-phosphate, sodium 2r,3s,5r-3-hydroxy-5-6-hydroxy-9h-purin-9-yl tetrahydrofuran-2-yl methyl phosphate |
| PubChem CID | 56776971 |
| SMILES | [Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=NC2=C1N=CNC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6427103
|
Thermo Scientific Chemicals
J6427103 |
1 g |
Each for $175.48
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14999-52-1 | |
| 376.17 | |
| VDPLPYMIJCLJEF-DZVPOJRENA-L | |
| 56776971 |
| C10H11N4Na2O7P | |
| MFCD00134874 | |
| sodium 2r,3s,5r-3-hydroxy-5-6-oxo-1h-purin-9 6h-yl tetrahydrofuran-2-yl methyl phosphate, 2'-deoxyinosine-5'-monophosphate, disodium salt, 2'-deoxyinosine 5'-monophosphate disodium salt, 2'-deoxyinosine-5'-monophosphate disodium salt, disodium dimp, 2'-deoxyinosine-5'-phosphate sodium, disodium 2'-deoxyinosine 5'-phosphate, sodium 2r,3s,5r-3-hydroxy-5-6-hydroxy-9h-purin-9-yl tetrahydrofuran-2-yl methyl phosphate | |
| [Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=NC2=C1N=CNC2=O |
Specifications
| 14999-52-1 | |
| C10H11N4Na2O7P | |
| 1 g | |
| VDPLPYMIJCLJEF-DZVPOJRENA-L | |
| disodium;[(2R,3S,5R)-3-hydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl phosphate | |
| 56776971 | |
| 99% | |
| 2'-Deoxyinosine-5'-monophosphate disodium salt |
| White | |
| MFCD00134874 | |
| sodium 2r,3s,5r-3-hydroxy-5-6-oxo-1h-purin-9 6h-yl tetrahydrofuran-2-yl methyl phosphate, 2'-deoxyinosine-5'-monophosphate, disodium salt, 2'-deoxyinosine 5'-monophosphate disodium salt, 2'-deoxyinosine-5'-monophosphate disodium salt, disodium dimp, 2'-deoxyinosine-5'-phosphate sodium, disodium 2'-deoxyinosine 5'-phosphate, sodium 2r,3s,5r-3-hydroxy-5-6-hydroxy-9h-purin-9-yl tetrahydrofuran-2-yl methyl phosphate | |
| [Na+].[Na+].O[C@H]1C[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=NC2=C1N=CNC2=O | |
| 376.17 | |
| 376.17 | |
| Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 239-090-3
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only