missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Deoxyguanosine Hydrate 99.0+%, TCI America™
Supplier: TCI America D005225G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 961-07-9 | |
| 267.25 | |
| YKBGVTZYEHREMT-DWFKHXNSNA-N | |
| 187790 | |
| 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one |
| C10H13N5O4 | |
| MFCD00150760 | |
| 2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl | |
| CHEBI:17172 | |
| NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1 |
Specifications
| 2′-Deoxyguanosine Hydrate | |
| White-Yellow | |
| MFCD00150760 | |
| 2'-deoxyguanosine, deoxyguanosine, guanine deoxyriboside, 2-deoxyguanosine, guanosine, 2'-deoxy, guanine deoxy nucleoside, 2'-deoxy-guanosine, 2;-deoxyguanosine, unii-g9481n71ro, guanine, 9-2-deoxy-beta-d-erythro-pentofuranosyl | |
| NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1 | |
| 267.25 | |
| CHEBI:17172 | |
| ≥99.0% (HPLC) |
| 961-07-9 | |
| C10H13N5O4 | |
| 25 g | |
| YKBGVTZYEHREMT-DWFKHXNSNA-N | |
| 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one | |
| 187790 | |
| 267.25 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6830&(8)-0681
RTECSNumber : MF8760000
TSCA : Yes