missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2'-Deoxyguanosine 5'-monophosphate, sodium salt hydrate, 99%
CAS: 52558-16-4 | C10H12N5Na2O7P | 391.19 g/mol
$228.70 - $228.70
Chemical Identifiers
| CAS | 52558-16-4 |
|---|---|
| Molecular Formula | C10H12N5Na2O7P |
| Molecular Weight (g/mol) | 391.19 |
| MDL Number | MFCD00150780 |
| InChI Key | CTPAMSRBXKGZCJ-WJQTVUBHNA-L |
| Synonym | deoxyguanosine monophosphate tetrahydrate sodium salt |
| PubChem CID | 131854716 |
| IUPAC Name | [(2R,3S,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium;tetrahydrate |
| SMILES | [Na+].[Na+].NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC226340010
|
Thermo Scientific Chemicals
226340010 |
1 g | Glass bottle |
Each for $228.70
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 52558-16-4 | |
| 391.19 | |
| CTPAMSRBXKGZCJ-WJQTVUBHNA-L | |
| 131854716 | |
| [Na+].[Na+].NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)N1 |
| C10H12N5Na2O7P | |
| MFCD00150780 | |
| deoxyguanosine monophosphate tetrahydrate sodium salt | |
| [(2R,3S,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium;tetrahydrate |
Specifications
| 52558-16-4 | |
| 0.65 to 0.68 (A280/A260) c=1,water,0.25 to 0.28 (A290/A260) c=1,water | |
| >245°C | |
| Authentic | |
| Glass bottle | |
| MFCD00150780 | |
| deoxyguanosine monophosphate tetrahydrate sodium salt | |
| [Na+].[Na+].NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)N1 | |
| 391.19 | |
| 391.19 | |
| Crystalline Powder |
| 98.5 | |
| 100.0 | |
| White | |
| 98.5% min. (HPLC) | |
| C10H12N5Na2O7P | |
| 1 g | |
| CTPAMSRBXKGZCJ-WJQTVUBHNA-L | |
| [(2R,3S,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium;tetrahydrate | |
| 131854716 | |
| 99% | |
| 2′-Deoxyguanosine 5′-monophosphate,sodium salt hydrate, 99% |
Safety and Handling
TSCA : TSCA
RUO – Research Use Only