missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Deoxycytidine 98.0+%, TCI America™
$53.16 - $159.49
Chemical Identifiers
| CAS | 951-77-9 |
|---|---|
| Molecular Formula | C9H13N3O4 |
| Molecular Weight (g/mol) | 227.22 |
| MDL Number | MFCD00006547 |
| InChI Key | CKTSBUTUHBMZGZ-JORGKRSHNA-N |
| Synonym | 2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine |
| PubChem CID | 13711 |
| ChEBI | CHEBI:15698 |
| IUPAC Name | 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one |
| SMILES | NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 |
Chemical Identifiers
| 951-77-9 | |
| 227.22 | |
| CKTSBUTUHBMZGZ-JORGKRSHNA-N | |
| 13711 | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one |
| C9H13N3O4 | |
| MFCD00006547 | |
| 2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine | |
| CHEBI:15698 | |
| NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 |
Specifications
| 951-77-9 | |
| White | |
| MFCD00006547 | |
| 2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine | |
| NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 | |
| 227.22 | |
| CHEBI:15698 | |
| ≥98.0% (HPLC,T) | |
| 2′-Deoxycytidine |
| 204°C | |
| C9H13N3O4 | |
| 1 g | |
| CKTSBUTUHBMZGZ-JORGKRSHNA-N | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one | |
| 13711 | |
| 227.22 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6834&(8)-0682
RTECSNumber : HA3800000
TSCA : Yes