missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxycytidine, 99+%

Catalog No. AC226720250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226720250 25 g Glass bottle
AC226720010 1 g Glass bottle
AC226720050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226720250 Supplier Thermo Scientific Chemicals Supplier No. 226720250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 951-77-9 | C9H13N3O4 | 227.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 951-77-9
Molecular Formula C9H13N3O4
Molecular Weight (g/mol) 227.22
MDL Number MFCD00006547
InChI Key CKTSBUTUHBMZGZ-JORGKRSHNA-N
Synonym 2'-deoxycytidine, deoxycytidine, dcyd, cytosine deoxyriboside, cytidine, 2'-deoxy, deoxyribose cytidine, desoxycytidin german, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidin-2 1h-one, cytosine, deoxyribonucleoside, d-cytidine
PubChem CID 13711
ChEBI CHEBI:15698
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 209°C to 211°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 227.22
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2'-Deoxycytidine
Show More Show Less
EINECSNumber 213-454-1

RUO – Research Use Only