missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxyadenosine Monohydrate 99.0+%, TCI America™

Supplier:  TCI America D004625G

Quantity 25 g

 View more versions of this product

Catalog No. D004625G


Only null left

Description

Description

Specifications

Chemical Identifiers

16373-93-6
251.25
OLXZPDWKRNYJJZ-YUZWJPFSNA-N
9549172
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
C10H13N5O3
MFCD00149364
2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate
(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol

Specifications

2′-Deoxyadenosine Monohydrate
189°C
C10H13N5O3
MFCD00149364
OLXZPDWKRNYJJZ-YUZWJPFSNA-N
(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
9549172
≥99.0% (T)
16373-93-6
White
25 g
2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
251.25
251.25
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6829&(8)-0683

RTECSNumber : AU7358600

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions