Learn More
Thermo Scientific Chemicals 2'-Deoxyadenosine monohydrate, 99%
CAS: 16373-93-6 | C10H13N5O3 | 251.25 g/mol
Supplier: Thermo Scientific Chemicals A1116606
| Quantity | 5 g |
|---|
Description
2?-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2?-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 16373-93-6 | |
| 251.25 | |
| OLXZPDWKRNYJJZ-YUZWJPFSNA-N | |
| 9549172 | |
| NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 |
| C10H13N5O3 | |
| MFCD00149364 | |
| 2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate | |
| (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol;hydrate |
Specifications
| 2'-Deoxyadenosine monohydrate | |
| 186°C to 189°C | |
| 5 g | |
| 5191174 | |
| Soluble in water. | |
| NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 | |
| (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol;hydrate | |
| 9549172 | |
| 99% |
| 16373-93-6 | |
| C10H13N5O3 | |
| MFCD00149364 | |
| 2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate | |
| OLXZPDWKRNYJJZ-YUZWJPFSNA-N | |
| −25° (c=0.5 water) | |
| 251.25 | |
| 269.26 (251.24 Anhydrous) |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 213-488-7
RTECSNumber : AU7358600
TSCA : Yes
Recommended Storage : Keep cold