missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyadenosine hydrate, 99%

Catalog No. AC154990250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154990250 25 g Glass bottle
AC154990010 1 g Glass bottle
AC154990050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC154990250 Supplier Thermo Scientific Chemicals Supplier No. 154990250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16373-93-6 | C10H13N5O3 | 251.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16373-93-6
Molecular Formula C10H13N5O3
Molecular Weight (g/mol) 251.25
MDL Number MFCD00149364
InChI Key OLXZPDWKRNYJJZ-YUZWJPFSNA-N
Synonym 2'-deoxyadenosine monohydrate, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol hydrate, adenosine, 2'-deoxy-, monohydrate, 2'-deoxyadenosine hydrate, 2-deoxyadenosine hydrate, deoxyadenosine hydrate, 9-2-deoxy-, a-d-ribofuranosyl adenine, deoxyadenosine monohydrate, 2-deoxyadenosine monohydrate, 2?-deoxyadenosine monohydrate
PubChem CID 9549172
SMILES NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 185.0°C to 187.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: 0.3% in water (20°C). Other solubilities: soluble in dmf,formamide,hot dioxane,methanol,ethanol,insoluble in hydrocarbons
IUPAC Name 5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Formula Weight 251.24
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxyadenosine hydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only