missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2'-Deoxyadenosine, 99%

CAS: 958-09-8 | C10H13N5O3 | 251.25 g/mol

Supplier:  Thermo Scientific Chemicals J6388618

Quantity 50 g

 View more versions of this product

Catalog No. AAJ6388618


Only null left

Description

Description

2'-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.

Solubility
Soluble in water and caustic soda.

Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

958-09-8
251.25
OLXZPDWKRNYJJZ-RRKCRQDMSA-N
13730
(2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
C10H13N5O3
MFCD00005754
2'-deoxyadenosine, deoxyadenosine, 2-deoxyadenosine, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, adenyldeoxyriboside, adenine deoxyribonucleoside, adenine deoxyribose, desoxyadenosine, ccris 1782, unii-p582c98ulc
CHEBI:17256
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1

Specifications

2'-Deoxyadenosine
White
MFCD00005754
2'-deoxyadenosine, deoxyadenosine, 2-deoxyadenosine, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, adenyldeoxyriboside, adenine deoxyribonucleoside, adenine deoxyribose, desoxyadenosine, ccris 1782, unii-p582c98ulc
OLXZPDWKRNYJJZ-RRKCRQDMSA-N
−25° (c=0.5 in Water)
251.25
CHEBI:17256
99%
958-09-8
C10H13N5O3
50 g
Soluble in water and caustic soda.
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
(2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
13730
251.24
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 213-488-7

RTECSNumber : AU7358650

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions