Learn More
Thermo Scientific Chemicals 2'-Deoxyadenosine, 99%
CAS: 958-09-8 | C10H13N5O3 | 251.25 g/mol
Supplier: Thermo Scientific Chemicals J6388614
| Quantity | 25 g |
|---|
Description
2'-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.
Solubility
Soluble in water and caustic soda.
Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 958-09-8 | |
| 251.25 | |
| OLXZPDWKRNYJJZ-RRKCRQDMSA-N | |
| 13730 | |
| (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol |
| C10H13N5O3 | |
| MFCD00005754 | |
| 2'-deoxyadenosine, deoxyadenosine, 2-deoxyadenosine, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, adenyldeoxyriboside, adenine deoxyribonucleoside, adenine deoxyribose, desoxyadenosine, ccris 1782, unii-p582c98ulc | |
| CHEBI:17256 | |
| NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 |
Specifications
| 2'-Deoxyadenosine | |
| White | |
| MFCD00005754 | |
| 2'-deoxyadenosine, deoxyadenosine, 2-deoxyadenosine, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, adenyldeoxyriboside, adenine deoxyribonucleoside, adenine deoxyribose, desoxyadenosine, ccris 1782, unii-p582c98ulc | |
| OLXZPDWKRNYJJZ-RRKCRQDMSA-N | |
| −25° (c=0.5 in Water) | |
| 251.25 | |
| CHEBI:17256 | |
| 99% |
| 958-09-8 | |
| C10H13N5O3 | |
| 25 g | |
| Soluble in water and caustic soda. | |
| NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 | |
| (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol | |
| 13730 | |
| 251.24 | |
| Powder |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 213-488-7
RTECSNumber : AU7358650
TSCA : Yes
Recommended Storage : Ambient temperatures