missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-D-glucose 97.0+%, TCI America™

Catalog No. D00515G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D00515G 5 g
D00511G 1 g
D005125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D00515G Supplier TCI America Supplier No. D00515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 154-17-6
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00151328
InChI Key PMMURAAUARKVCB-PHUJZJCSNA-N
Synonym deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose
PubChem CID 17751002
IUPAC Name (4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O

Specifications

Melting Point 150°C
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Deoxy-D-glucose
RTECSNumber MQ3325000
TSCA Yes