missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Deoxy-D-glucose 97.0+%, TCI America™
Supplier: TCI America D005125G
| Quantity | 25 g |
|---|
Chemical Identifiers
| C6H12O5 | |
| MFCD00151328 | |
| deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose | |
| (4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol |
Specifications
| 2-Deoxy-D-glucose | |
| 150°C | |
| 25 g | |
| deoxyglucose, 2-deoxy-d-mannose, 2-deoxy-d-arabinohexose, unii-9g2mp84a8w, d-arabino-hexose, 2-deoxy, arabino-hexose, 2-deoxy, d-glucose, 2-deoxy, 3r,4s,5r-3,4,5,6-tetrahydroxyhexanal, 2 deoxyglucose, 2 deoxy d glucose | |
| OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O | |
| 164.16 | |
| 164.16 | |
| Crystalline Powder |
| 154-17-6 | |
| C6H12O5 | |
| MFCD00151328 | |
| PMMURAAUARKVCB-PHUJZJCSNA-N | |
| (4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol | |
| 17751002 | |
| ≥97.0% (GC) |
Safety and Handling
RTECSNumber : MQ3325000
TSCA : Yes