missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Deoxy-D-galactose, 99%

Catalog No. ac111970010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111970010 1 g Glass bottle
AC111970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111970010 Supplier Thermo Scientific Chemicals Supplier No. 111970010
Only null left

CAS: 1949-89-9 | C6H12O5 | 164.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1949-89-9
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00014649
InChI Key PMMURAAUARKVCB-ONLQHTGVNA-N
Synonym 2-deoxy-d-galactose, 3r,4r,5r-3,4,5,6-tetrahydroxyhexanal, d-lyxo-hexose, 2-deoxy, unii-531k2iok5q, 2-deoxy-d-lyxo-hexose, lyxo-hexose, 2-deoxy, ksc179c4l
PubChem CID 102191
ChEBI CHEBI:27411
IUPAC Name (3R,4R,5R)-3,4,5,6-tetrahydroxyhexanal
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 109.0°C to 111.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation + 57.00
Formula Weight 164.16
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Deoxy-D-galactose
Show More Show Less
EINECSNumber 217-765-3

RUO – Research Use Only