missing translation for 'onlineSavingsMsg'
Learn More

2'-Deoxy-2'-fluorouridine 97.0+%, TCI America™

Catalog No. D36155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36155G 5 g
D36151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36155G Supplier TCI America Supplier No. D36155G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 784-71-4
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD01317293
InChI Key UIYWFOZZIZEEKJ-XVFCMESISA-N
Synonym 2'-deoxy-2'-fluorouridine, 2'-fluoro-2'-deoxyuridine, unii-y2yc903qw8, uridine, 2'-deoxy-2'-fluoro, 2'-deoxy-2'-fluoro-uridine, 2-fluoro-2-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4-dione, 2'fluoro-2'-deoxyuridine, 1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl-3h-pyrimidine-2,4-dione, pubchem14203
PubChem CID 150851
IUPAC Name 1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H]([C@H](F)[C@@H]1O)N1C=CC(=O)NC1=O

Specifications

Melting Point 151°C
Color White
Quantity 5 g
Formula Weight 246.19
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Deoxy-2′-fluorouridine