missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-2-fluoro-1,3,5-tri-O-benzoyl-alpha-D-arabinofuranose 98.0+%, TCI America™

Catalog No. p-7128357
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45941G 1 g
D45945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45941G Supplier TCI America Supplier No. D45941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 97614-43-2
Molecular Formula C26H21FO7
Molecular Weight (g/mol) 464.445
MDL Number MFCD00083339
InChI Key JOAHVPNLVYCSAN-UXGLMHHASA-N
Synonym 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-arabinofuranose, 2r,3s,4r,5r-5-benzoyloxy methyl-3-fluorotetrahydrofuran-2,4-diyl dibenzoate, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-, a-d-arabinofuranose, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-a-d-arabinofuranose, 2r,3r,4s,5r-3,5-bis benzoyloxy-4-fluorooxolan-2-yl methyl benzoate, 2r,3s,4r,5r-5-benzoyloxymethyl-3-fluoro-tetrahydrofuran-2,4-diyl dibenzoate, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-md-arabinose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-d-ribofuranose, pubchem20401, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-ribofuranose
PubChem CID 11754171
IUPAC Name [(2R,3R,4S,5R)-3,5-dibenzoyloxy-4-fluorooxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)F)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 85°C
Color White
Quantity 1 g
Formula Weight 464.45
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Deoxy-2-fluoro-1,3,5-tri-O-benzoyl-alpha-D-arabinofuranose