missing translation for 'onlineSavingsMsg'
Learn More

2-Decylthiophene 97.0+%, TCI America™

Catalog No. D52405ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52405ML 5 mL
D52401ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52405ML Supplier TCI America Supplier No. D52405ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24769-39-9
Molecular Formula C14H24S
Molecular Weight (g/mol) 224.406
MDL Number MFCD00715761
InChI Key GOTHKCARNFTUSW-UHFFFAOYSA-N
PubChem CID 566331
IUPAC Name 2-decylthiophene
SMILES CCCCCCCCCCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 167°C
Quantity 5 mL
Formula Weight 224.41
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Decylthiophene