missing translation for 'onlineSavingsMsg'
Learn More

2-Decanol 98.0+%, TCI America™

Supplier:  TCI America D137925ML

Quantity 25 mL

 View more versions of this product

Catalog No. D137925ML


Only null left

Description

Description

Specifications

Chemical Identifiers

1120-06-5
158.285
ACUZDYFTRHEKOS-UHFFFAOYSA-N
14254
CCCCCCCCC(C)O
C10H22O
MFCD00004594
2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
decan-2-ol

Specifications

2-Decanol
-4°C
211°C
25 mL
2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
CCCCCCCCC(C)O
158.285
158.29
Liquid
1120-06-5
Colorless
C10H22O
MFCD00004594
ACUZDYFTRHEKOS-UHFFFAOYSA-N
decan-2-ol
14254
≥98.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions