missing translation for 'onlineSavingsMsg'
Learn More

2-Decanol, 98%

CAS: 1120-06-5 | C10H22O | 158.285 g/mol

Supplier:  Thermo Scientific Chemicals L0568814

 View more versions of this product

Catalog No. AAL0568814


Only null left

Description

Description

2-Decanol is used to develop a miniature catalytic reactor for the oxidation of alcohols with O2 in supercritical CO2. It was used to study the substrate spectrum of phytanoyl-CoA hydroxylase with regard to the length of both the acyl chain and the branch at position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Decanol is used to develop a miniature catalytic reactor for the oxidation of alcohols with O2 in supercritical CO2. It was used to study the substrate spectrum of phytanoyl-CoA hydroxylase with regard to the length of both the acyl chain and the branch at position.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in well sealed container.
Specifications

Specifications

2-Decanol
-5°C
211°C
C10H22O
MFCD00004594
1719664
Not miscible in water.
CCCCCCCCC(C)O
158.285
158.29
1120-06-5
0.825
85°C (185°F)
1.434
25 g
2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
ACUZDYFTRHEKOS-UHFFFAOYSA-N
decan-2-ol
14254
98%

Chemical Identifiers

1120-06-5
158.285
ACUZDYFTRHEKOS-UHFFFAOYSA-N
14254
CCCCCCCCC(C)O
C10H22O
MFCD00004594
2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
decan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 214-296-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only