missing translation for 'onlineSavingsMsg'
Learn More

2-Decanol, 98%

Catalog No. AAL0568806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0568806 5 g
AAL0568814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0568806 Supplier Thermo Scientific Chemicals Supplier No. L0568806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1120-06-5 | C10H22O | 158.285 g/mol

2-Decanol is used to develop a miniature catalytic reactor for the oxidation of alcohols with O2 in supercritical CO2. It was used to study the substrate spectrum of phytanoyl-CoA hydroxylase with regard to the length of both the acyl chain and the branch at position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Decanol is used to develop a miniature catalytic reactor for the oxidation of alcohols with O2 in supercritical CO2. It was used to study the substrate spectrum of phytanoyl-CoA hydroxylase with regard to the length of both the acyl chain and the branch at position.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 1120-06-5
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00004594
InChI Key ACUZDYFTRHEKOS-UHFFFAOYSA-N
Synonym 2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
PubChem CID 14254
IUPAC Name decan-2-ol
SMILES CCCCCCCCC(C)O

Specifications

Melting Point -5°C
Density 0.825
Boiling Point 211°C
Flash Point 85°C (185°F)
Refractive Index 1.434
Quantity 5 g
Beilstein 1719664
Solubility Information Not miscible in water.
Formula Weight 158.29
Percent Purity 98%
Chemical Name or Material 2-Decanol
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 214-296-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only