missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Decanol, 98%
CAS: 1120-06-5 | C10H22O | 158.28 g/mol
$157.10
Chemical Identifiers
| CAS | 1120-06-5 |
|---|---|
| Molecular Formula | C10H22O |
| Molecular Weight (g/mol) | 158.28 |
| MDL Number | MFCD00004594 |
| InChI Key | ACUZDYFTRHEKOS-UHFFFAOYSA-N |
| Synonym | 2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477 |
| PubChem CID | 14254 |
| IUPAC Name | decan-2-ol |
| SMILES | CCCCCCCCC(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC331600250
|
N/A | 25 g |
Each for $157.10
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C10H22O | |
| MFCD00004594 | |
| 2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477 | |
| decan-2-ol |
Specifications
| 1120-06-5 | |
| 100.0 | |
| Colorless | |
| 211°C | |
| Authentic | |
| Glass bottle | |
| 1.4330 to 1.435 | |
| MFCD00004594 | |
| 0.82 | |
| Solubility in water: insoluble in water | |
| CCCCCCCCC(C)O | |
| 158.28 | |
| 158.28 | |
| Liquid |
| 97.5 | |
| -5°C | |
| 0.8200g/mL | |
| 85°C | |
| 97.5% min. (GC) | |
| C10H22O | |
| CH3(CH2)7CH(OH)CH3 | |
| 01, I, 213 | |
| 2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477 | |
| ACUZDYFTRHEKOS-UHFFFAOYSA-N | |
| decan-2-ol | |
| 14254 | |
| 98% | |
| 2-Decanol, 95% |
Safety and Handling
EINECSNumber : 214-296-6
TSCA : TSCA
RUO – Research Use Only