missing translation for 'onlineSavingsMsg'
Learn More

2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde, 97%, Thermo Scientific™

Catalog No. p-7033081
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5009103 1g
AAH5009106 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5009103 Supplier Thermo Scientific Chemicals Supplier No. H5009103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 902835-88-5
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.203
MDL Number MFCD07781136
InChI Key AEOXNPSSZSDLDK-UHFFFAOYSA-N
Synonym 2-cyclopropylmethoxy nicotinaldehyde, 2-cyclopropylmethoxy pyridine-3-carbaldehyde, 2-cyclopropylmethoxy pyridine-3-carboxaldehyde, 2-cyclopropylmethoxy-pyridine-3-carbaldehyde, 2-cyclopropyl-methoxy pyridine-3-carbaldehyde
PubChem CID 42553129
IUPAC Name 2-(cyclopropylmethoxy)pyridine-3-carbaldehyde
SMILES C1CC1COC2=C(C=CC=N2)C=O

Specifications

Quantity 1g
Formula Weight 177.2
Percent Purity 97%
Chemical Name or Material 2-(Cyclopropylmethoxy)pyridine-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only