missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™

Catalog No. C22761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22761G 1 g
C22765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22761G Supplier TCI America Supplier No. C22761G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 126689-01-8
Molecular Formula C9H17BO2
Molecular Weight (g/mol) 168.04
MDL Number MFCD05663847
InChI Key XGBMQBPLWXTEPM-UHFFFAOYSA-N
Synonym cyclopropylboronic acid pinacol ester, cyclopropylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclopropane, 1,3,2-dioxaborolane, 2-cyclopropyl-4,4,5,5-tetramethyl, 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborol, cypbe, pubchem4015, pubchem7931, acmc-209bbh
PubChem CID 2758015
IUPAC Name 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1CC1

Specifications

Color Yellow
Boiling Point 146°C
Quantity 1 g
UN Number 3272
Formula Weight 168.04
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane