missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentylcyclopentanone 99.0+%, TCI America™

Catalog No. C093125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C093125ML 25 mL
C0931500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C093125ML Supplier TCI America Supplier No. C093125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4884-24-6
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00051419
InChI Key CWZGKTMWPFTJCS-UHFFFAOYNA-N
Synonym 2-cyclopentylcyclopentanone, 1,1'-bicyclopentyl-2-one, bicyclopentyl-2-one, 1,1'-bi cyclopentan-2-one, bicyclopentylone, acmc-1aqra, 2-cyclopentyl cyclopentanone, cyclopentanone, 2-cyclopentyl
PubChem CID 21003
IUPAC Name [1,1'-bi(cyclopentane)]-2-one
SMILES O=C1CCCC1C1CCCC1

Specifications

Color Yellow
Boiling Point 233°C
Quantity 25 mL
Formula Weight 152.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclopentylcyclopentanone
EINECSNumber (4)-1368
TSCA Yes