missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentene-1-acetic acid, tech. 90%, remainder mainly 3-cyclopentene-1-acetic acid

Catalog No. AAH2750706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2750706 5 g
AAH2750714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2750706 Supplier Thermo Scientific Chemicals Supplier No. H2750706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13668-61-6 | C7H10O2 | 126.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13668-61-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00001400
InChI Key NNRZTJAACCRFRV-UHFFFAOYSA-N
Synonym 2-cyclopentene-1-acetic acid, 2-cyclopent-2-en-1-yl acetic acid, cyclopent-2-enylacetic acid, 2-cyclopenten-1-ylacetic acid, cyclopent-2-en-1-ylacetic acid, cyclopentene-3-acetic acid, 2-cyclopentene-1-aceticacid, acmc-1c47k, 2-cyclopent-2-enylacetic acid, cyclopent-2-ene-1-acetic acid
PubChem CID 96216
IUPAC Name 2-cyclopent-2-en-1-ylacetic acid
SMILES C1CC(C=C1)CC(=O)O

Specifications

Melting Point 19°C
Density 1.047
Boiling Point 238°C
Flash Point 143°C (289°F)
Odor Rancid
Refractive Index 1.468
Quantity 5 g
Beilstein 2042172
Formula Weight 126.15
Percent Purity 90%
Grade Technical
Assay remainder mainly 3-cyclopentene-1-acetic acid
Chemical Name or Material 2-Cyclopentene-1-acetic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-146-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only