missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopenten-1-one 96.0+%, TCI America™

Catalog No. C07945ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07945ML 5 mL
C079425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07945ML Supplier TCI America Supplier No. C07945ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 930-30-3
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.10
MDL Number MFCD00001401
InChI Key BZKFMUIJRXWWQK-UHFFFAOYSA-N
Synonym 2-cyclopenten-1-one, 2-cyclopentenone, cyclopentenone, cyclopent-2-enone, cyclopenten-3-one, 2-cyclopentene-1-one, cyclopent-2-ene-1-one, unii-q0u2igf9ck, 3-cyclopenten-2-one, q0u2igf9ck
PubChem CID 13588
IUPAC Name cyclopent-2-en-1-one
SMILES O=C1CCC=C1

Specifications

Color Yellow
Boiling Point 154°C
Quantity 5 mL
UN Number 1224
Formula Weight 82.10
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Cyclopenten-1-one
EINECSNumber (3)-4493
TSCA No
Recommended Storage Refrigerator