missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethylboronic acid pinacol ester, 96%

Catalog No. AAH3115606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3115606 5 g
AAH3115603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3115606 Supplier Thermo Scientific Chemicals Supplier No. H3115606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 167692-95-7 | C14H27BO2 | 238.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 167692-95-7
Molecular Formula C14H27BO2
Molecular Weight (g/mol) 238.178
MDL Number MFCD12546193
InChI Key LDKDGYZAIIKCGK-UHFFFAOYSA-N
Synonym 2-cyclohexylethylboronic acid pinacol ester, 2-2-cyclohexylethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-cyclohexylethylboronicacid
PubChem CID 15259990
IUPAC Name 2-(2-cyclohexylethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2CCCCC2

Specifications

Quantity 5 g
Formula Weight 238.18
Percent Purity 96%
Chemical Name or Material 2-Cyclohexylethylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only