missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethylamine 98.0+%, TCI America™

Catalog No. C27651G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27651G 1 g
C27655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27651G Supplier TCI America Supplier No. C27651G

Chemical Identifiers

CAS 4442-85-7
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00058668
InChI Key HFACYWDPMNWMIW-UHFFFAOYSA-N
Synonym 2-cyclohexylethylamine, 2-cyclohexyl-ethylamine, cyclohexaneethanamine, cyclohexaneethylamine, 2-cyclohexylethan-1-amine, cyclohexaneethanamine 9ci, 2-aminoethylcyclohexane, ethylamine, 2-cyclohexyl, chembl19428, 2-cyclohexylethyl amine hydrochloride
PubChem CID 20509
IUPAC Name 2-cyclohexylethan-1-amine
SMILES NCCC1CCCCC1

Specifications

Color Colorless
Boiling Point 96°C
Quantity 1 g
UN Number 2733
Formula Weight 127.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Cyclohexylethylamine
RTECSNumber KR3325000
TSCA No