missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethylamine, 97%

Catalog No. p-7030335
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2798703 1 g
AAH2798706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2798703 Supplier Thermo Scientific Chemicals Supplier No. H2798703

CAS: 4442-85-7 | C8H17N | 127.23 g/mol

In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Protect from direct light.

Chemical Identifiers

CAS 4442-85-7
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00058668
InChI Key HFACYWDPMNWMIW-UHFFFAOYSA-N
Synonym 2-cyclohexylethylamine, 2-cyclohexyl-ethylamine, cyclohexaneethanamine, cyclohexaneethylamine, 2-cyclohexylethan-1-amine, cyclohexaneethanamine 9ci, 2-aminoethylcyclohexane, ethylamine, 2-cyclohexyl, chembl19428, 2-cyclohexylethyl amine hydrochloride
PubChem CID 20509
IUPAC Name 2-cyclohexylethanamine
SMILES NCCC1CCCCC1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air sensitive
Formula Weight 127.23
Percent Purity 97%
Chemical Name or Material 2-Cyclohexylethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 224-673-7
RTECSNumber KR3325000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only