missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethyl-4-O-(alpha-D-glucopyranosyl)-beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Catalog No. aaj66517md
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66517MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66517MD Supplier Thermo Scientific Chemicals Supplier No. J66517MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 260804-65-7 | Molecular Formula: C20H36O11 | Molecular Weight: 452.497 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 260804-65-7
Molecular Formula C20H36O11
Molecular Weight (g/mol) 452.497
MDL Number MFCD01862984
InChI Key RHXPDNGQJSXOMW-OIIXUNCGSA-N
Synonym 2-cyclohexylethyl-4-o-a-d-glucopyranosyl-b-d-glucopyranoside, ma5, 2-6-2-cyclohexylethoxy-tetrahydro-4,5-dihydroxy-2 hydroxymethyl-2h-pyran-3-yloxy-tetrahydro-6 hydroxymethyl-2h-pyran-3,4,5-triol, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-6-2-cyclohexylethoxy-4,5-dihydroxy-2-hydroxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, cymal-2, cymal™-2, cymal r-2, 2-cyclohexylethyl beta-maltoside, 2-cyclohexylethyl, a-d-maltoside, 2-cyclohexylethyl-beta-d-maltoside
PubChem CID 9547906
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-(2-cyclohexylethoxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1CCC(CC1)CCOC2C(C(C(C(O2)CO)OC3C(C(C(C(O3)CO)O)O)O)O)O

Specifications

Color White
Quantity 250 mg
Sensitivity Moisture sensitive
Formula Weight 452.49
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Cyclohexylethyl-4-O-(alpha-D-glucopyranosyl)-beta-D-glucopyranoside
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only